Slide-Rite™ Bag, Uline
Supplier: ULINE
S-10837
76241-136CS 314.03 USD
76241-136
Slide-Rite™ Bag, Uline
Bags Storage Bags
This bag protects items from dirt and moisture.
  • Freezer safe, acid free and archival safe
  • 100% virgin polyethylene film
This bag guarantee to produce a durable, airtight seal every time.
Certifications: FDA and USDA compliant for food contact.