Micro-Lite® High Intensity LED Ring Illuminator, O.C. White
Supplier: O. C. WHITE CO.
LV2000-B
76241-014EA 1455.65 USD
76241-014 76247-642
Micro-Lite® High Intensity LED Ring Illuminator, O.C. White
Lights LED Lights, Lamps and Bulbs
This LED illuminator is the brightest, and rivals 150 W halogen illuminators with the same yellowish light color.
  • Total energy consumption of this unit is a scant 8 W
  • Features auto switchable transformer design
  • Black molded shades, ESD safe