Super-Tack™ Construction Tape, Ultratape Industries
Supplier: Ultratape Industries
1160WH100-P3D
76240-852CS 1728.3 USD
76240-852
Super-Tack™ Construction Tape, Ultratape Industries
Tapes
UltraTape Construction tapes are designed for critical environment construction and support applications. They can be used for hanging heavy plastic sheeting to create temporary walls, taping cart wheels and ductwork, floor marking, and color coding. Use the colored, striped hazard warning tapes to designate the OSHA critical areas in cleanrooms.
  • Super-Tack™ construction tape has a strong adhesive bond that leaves no residue upon removal
  • Protocol™ tapes for immediate recognition that certified cleanroom compatible tape is in use
Delivery information: Available in UltraClean and standard clean.