6200 Series Half Facepiece Respirators, 3M™
Supplier: 3M
7000001933
76240-656CS 1357.39 USD
76240-656
6200 Series Half Facepiece Respirators, 3M™
Respirators Reusable Respirators Half Face Respirators
3M™ half facepiece reusable respirator 6200 offers reliable and convenient respiratory protection and is suitable for many situations, helping provide protection against particulates and a wide variety of gases.
  • Available in 3 sizes (small, medium, large) to comfortably fit many faces
  • Head harness assembly and spare parts available
When properly fitted and used with approved 3M™ cartridges and filters, helps provide respiratory protection from a variety of gases, vapors, and particulate hazards at concentrations up to 10 times the Permissible Exposure Limit (PEL).
Packaging: Comes with sealable/reclosable storage bag.
Caution: Not for use in environments that are immediately dangerous to life or health (IDLH).