CleanWIPE® Contamination Control Swabs, Foam Head, Conical Tip, Foamtec International WCC
Supplier: FOAMTEC INTERNATIONAL, LLC.
HT1032
76240-600CS 3215.04 USD
76240-600
CleanWIPE® Contamination Control Swabs, Foam Head, Conical Tip, Foamtec International WCC
Swabs Swabs for Cleaning
Designed to meet demanding cleanroom applications.
  • Features a 100 ppi polyurethane head and a polypropylene handle
  • Handle Ø: 2.5 mm
Can be used with alcohol, Freon® 113 fluorocarbon, acetone and other solvents for assembly cleaning, and for epoxy application.
Ordering information: Available in pyrogen-free versions upon request.
Packaging: Packaged in polybags.