Benchtop Air Ionizer, SCS, Desco Industries
Supplier: Desco
963E
76240-436EA 1753.17 USD
76240-436
Benchtop Air Ionizer, SCS, Desco Industries
Ionizers
The Benchtop Air Ionizer is a self-contained ionizing air blower designed to remove static charges from non-conductive objects.
  • Steady-state DC benchtop ionizer with auto-balance
  • Provides fast discharge with low airflow for greater operator comfort
  • Less than 2 seconds discharge time with ±15 volts offset voltage (balance)
  • Electrostatically shielded emitter points eliminate field-induced charging
  • No grounding wires required, ensuring quick and easy installation and setup
  • North American power adapter included
The proprietary circuitry contained in the blower generates balanced levels of positive and negative ions. The ionizer is equipped with a two speed fan which allows the user to select the amount of ionized air to be delivered to the target object. Ionization performance is consistent at all speeds and allows for use with sensitive applications, such as calibration, test, or repair procedures.
Certifications: Meets the ANSI/ESD S20.20 required limits for ionization tested per ANSI/ESD STM3.1 and ESD TR53. Certifications: UL, c-UL, NOM, CE.