Pegboard Panel, Arlink
Supplier: ARLINK DIVISION CUST SERV TE
ARL-8334
76240-260EA 303.31 USD
76240-260
Pegboard Panel, Arlink
Racks Draining Racks
Sturdy painted steel pegboard panel, ideal for hanging tools.
  • Mounts in micro slots
  • Adjusts vertically in 1" increments