Silbrade® Braid Reinforced Silicone Tubing, Newage Industries
Supplier: NewAge Industries
290-0457-100
76240-168EA 2729.83 USD
76240-168 76240-170
Silbrade® Braid Reinforced Silicone Tubing, Newage Industries
Tubing Transfer Tubing
Silbrade® Braid Reinforced Silicone Tubing offers excellent product protection, product visibility, and flexibility. It is used widely in the Food, Dairy, Beverage, Biotech, Pharmaceutical, and all other Sanitary Process industries to transport products and fluids.
  • Available in custom lengths
  • Open white mesh polyester braid
  • Silicone elastomer core and cover layer
Adhesive lines, beverage service, bottle filling, chemical feeds, deionized water transfer, fermenter lines, hot fill lines, food handling, laboratory, resin feeds, stirring vessels, viscous material handling.