Weller® Precision Tweezer with Curved Tips, Apex Tool Group
Supplier: Cooper Tools
51SA
76240-108EA 117.55 USD
76240-108
Weller® Precision Tweezer with Curved Tips, Apex Tool Group
Designed for confined spaces.
The special stainless steel is non-magnetic. The tweezers are non-rusting, acid-proof and heat resistant up to 300 °C (512 °F). Tweezers made from special stainless steel have the identification letter "SA" in their order numbers. For applications in biology, medicine, laboratory technology, and microelectronics.