Weller® Precision Tweezer with Flat Rounded Tips, Apex Tool Group
Supplier: Cooper Tools
2ASA
76240-102EA 72.85 USD
76240-102
Weller® Precision Tweezer with Flat Rounded Tips, Apex Tool Group
Forceps
Designed for gripping components.
The special stainless steel is non-magnetic. The tweezers are non-rusting, acid-proof and heat resistant up to 300 °C (512 °F). Tweezers made from special stainless steel have the identification letter "SA" in their order numbers. Suitable for all standard gripping applications and assembly jobs on printed-circuit boards.