SMD Tweezers, Lindstrom Precision Tools
Supplier: J.H. Williams Tool Group (Lindstrom)
SM111-SA
76240-098EA 96.84 USD
76240-098
SMD Tweezers, Lindstrom Precision Tools
Forceps
Used for positioning 5 mm monolithic chip capacitors. These tweezers are meant for quick and easy handling and positioning of any type of SMD (surface mount device) componenents.
Blunted edges prevent damage to printed circuit boards.