Adjustable Wrist Strap, SCS, Desco Industries
Supplier: Desco
2204
76240-048CS 656.46 USD
76240-048
Adjustable Wrist Strap, SCS, Desco Industries
Grounders Wrist Grounders
Adjustable wrist straps provide a comfortable, custom fit. One size adjusts to most any size wrist to provide reliable, 360° protection.
Certifications: Meets ANSI/ESD S20.20 and ANSI/ESD S1.1.