Clay Desiccants, Desiccare
Supplier: DESICCARE INC MS
02BM10R13
76239-974CS 109.18 USD
76239-974
Clay Desiccants, Desiccare
Desiccators Desiccants
This montmorillonite clay is a calcium aluminum silicate which is ground, processed and bagged.
  • Non-dusting for crucial applications
  • Highly effective in normal temperature and humidity ranges
  • Provides humidity control for sealed packages and containers
This desiccant is easily regenerated at low temperatures for repeated use.
Certifications: Meets or exceeds MIL-D3464 Type I and II specifications and approved by FDA for type III packaging.
Packaging: Packed in kraft paper pack which provides moisture proof.