Anti-Static Spray Bottles, All-Spec Industries
Supplier: HISCO, INC TE
SCB-32-ESD
76239-822CS 582.93 USD
76239-822
Anti-Static Spray Bottles, All-Spec Industries
Bottles Spray Bottles
ESD-safe spray polyethylene bottles with a concentrate that does not contain 'ethoxylated tertiary amine', which is widely used as an anti-static agent.
  • Average surface resistance of 10⁹ to 1010
  • Do not have the undesirable features associated with surfactant-laden material
The bottles dissipate a charge of ±5000 V in less than two seconds at 40% RH.