ESD Wash Bottle, R & R Lotion
Supplier: R & R Lotion
WHB-16-ESD
76239-634CS 1711.94 USD
76239-634
ESD Wash Bottle, R & R Lotion
Bottles Wash Bottles
This cleanroom blue static dissipative solvent containers have an average surface resistivity of 10⁹ to 10¹⁰.
  • No chemical reactivity with solvents such as TCE or alcohol
  • No color change or any other physical change noted for any solvents tested
  • Low density ESD safe with closure
  • 90 degree stem molded in one piece for leak proof service
The pH of the deionized water solutions tested was the same before and after a 7-day period.