Saturix Laundered, Heavyweight Polyester Knit Wipes, 2-Ply, Quilted
Supplier: FG CLEAN WIPES TE
7XCPP99X00
76239-462CS 1442.25 USD
76239-462
Saturix Laundered, Heavyweight Polyester Knit Wipes, 2-Ply, Quilted
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Heavyweight sealed edge polyester knit cleanroom wipes feature heavy-duty, 2-ply quilted construction for tough jobs in critical environments.
  • Ultra-pure 100% continuous filament polyester knit fabric construction (241 g/m²)
  • 2-ply substrate with quilted texture and laser sealed edges
  • Cleanliness and dependability for ISO Class 4 cleanroom use
  • Extremely durable and absorbent
  • Quilted surface pattern cleans more effectively and efficiently
  • Ultra-low particle generation, extractables, and NVRs
Meticulous attention to detail and sealed edges ensure ultra-low NVRs, extractables, and ions. The 2-ply construction traps and retains more liquid while the quilted surface texture removes surface contaminants with optimal efficiency. Wipes are laundered and packaged in an ISO Class 4 cleanroom using a proprietary laundering process and wash formulation.
Applications include large spill pick-ups, wiping and cleaning of abrasive surfaces, pre-sterilization product cleaning, as well as use in semiconductor and microelectronics manufacturing, and pharmaceutical and biotech manufacturing industries. Recommended for use in ISO Class 4 or higher environments.