Bio Barb® Adapters, Bioconnexx
Supplier: BIOCONNEXX INC. TE
PP-100-TC0375 PP-100-TC0500 PP-MINI-TC0500 PP-100-TC1000 PP150TC-0500HB PP-MINI-TC0250
76239-174EA 44.79 USD
76239-174 76239-176 76240-128 76240-130 76245-284 76246-022
Bio Barb® Adapters, Bioconnexx
Fittings and Connectors
True bore design allows for full flow and no restriction of product transfer. Eliminates turbulence caused by the adapter reducees possible damage to the medium being transferred.
  • Slight raise at the port on the TC face ensuring proper capture of gasket
  • Produced from a polypropylene resin
Certifications: Polypropylene complies with FDA and USP Class VI requirements.