Plastic Bio-Clamps, Bioconnexx
Supplier: BIOCONNEXX INC. TE
NG075WH NG150WH NG200WH NG300WH
76239-006EA 39.04 USD
76239-006 76239-008 76239-010 76239-012
Plastic Bio-Clamps, Bioconnexx
Clamps General Purpose Clamps
The Bio Clamp's design and construction lend itself to increased heat dissipation during high temperature applications.
  • Designed to fit sanitary flanges
  • Tamper evident
  • ISO Class 7 Clean Room produced and packaged
Reducing heat in non-metallic ferrules reduces the chance of warping and allowing leaks to occur. It can be sterilized by either steam or gamma irradiation.
Certifications: Conforms to ISO 2853.