Lindstrom RX Cutters
Supplier: J.H. Williams Tool Group (Lindstrom)
RX-8140 RX-8141 RX8142 RX-8150 RX8247 TRX-8180 RX-8147 RX8142-S RX-8148
76238-912EA 179.22 USD
76238-912 76238-914 76238-916 76238-918 76238-920 76238-922 76238-924 76238-930 76240-060 76246-000 89168-977
Lindstrom RX Cutters
Cutters
Lindstrom RX Cutters are made from high performance alloy steel.
  • Precision induction hardened edges 63–65 HRC
  • Cutting capacity is listed for solid copper wire
  • ESD safe handles in two-component synthetic material (Except 76238-930)
These cutters are developed according to the ERGO® process for a comfortable and effective grip in all situations. The presence of precision screw joint that minimizes friction and maximizes alignment of cutting edges.
Biospring® reduces tension throughout the working cycle of the tool and can be adjusted in three different positions.