Labx® 170 80% Abaca/20% Wood Pulp Nonwoven Cleanroom Wipers, Berkshire
Supplier: Berkshire
LB170.03D.32
76238-870CS 1016.09 USD
76238-870
Labx® 170 80% Abaca/20% Wood Pulp Nonwoven Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Labx® 170 is a cleaner alternative to industrial tissues and wipers.
  • Nonabrasive
  • Low extractables
  • High absorbency to weight ratio
Labx® 170 is an environmentally friendly wipe that has been designed for ISO Class 6 (Class 1000) and above cleanroom applications. These wipes are 100% biodegradable, and are made from 80% abaca and 20% wood pulp. Labx® 170 is specifically engineered as a lightweight and low lint material.
Certifications: ISO Class 6 to 8. Cleanroom Class 1000 to 100000