Saturix Laundered Polyester Knit Cleanroom Wipes, Sealed Edge
Supplier: FG CLEAN WIPES TE
73400SE99PS01
76238-852CS 1248.87 USD
76238-852
Saturix Laundered Polyester Knit Cleanroom Wipes, Sealed Edge
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Combining economy and cleanliness in one polyester knit wipe.
  • Polyester knit construction (145 g/m²)
  • Excellent balance of abrasion resistance and absorbancy
  • Assured cleanliness with low levels of ions and extractables
Laundered polyester knit wipes deliver the low particles, fibers, and NVRs of a dependable polyester knit, but are priced for value. With sealed edges and lightweight construction, these are optimal wipes for any environment at an everyday price point.
Applications include use in biotech, compounding pharmacies, electronics, medical device, or pharmaceutical industries. Recommended for use in ISO 4 - 8 controlled environments.