Saturix C30 Polyester/Cellulose Wipes
Supplier: FG CLEAN WIPES TE
7C30BBP04 7C3099P09
76238-842CS 696.7 USD
76238-842 76238-846
Saturix C30 Polyester/Cellulose Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These polyester/cellulose wipes offer the essential benefits of a non-woven wipe, high absorbency with aqueous liquids, dependable durability, and value. The exacting manufacturing process combines versatility, economy, and cleanliness.
  • Hydroentangled blend of 55% cellulose and 45% polyester fabric (64 g/m²)
  • Super absorbent
  • Extremely low particle generation
  • No chemical binders
Economical, general-purpose wipes offer extremely low particle generation with no chemical binders. Manufactured with a standard weight, hydroentangled blend of 55% cellulose and 45% polyester, C30 wipes are highly absorbent and residue-free, with low NVRs and ion extractalbes. Recommended for use in ISO 5 to 8 controlled environments.