Fiber-Free Face Masks, Foamtec
Supplier: Foamtec International
HT-4401
76238-802CS 2780.67 USD
76238-802
Fiber-Free Face Masks, Foamtec
Masks Hygiene Masks
These masks provide continuous particle/moisture control and cool, flexible comfort without the speech constraints, goggle/glasses steaming or moisture build-up on the face typical of paper and dense cloth masks.
  • Restricts vapor transfer from breath to work surface
  • Cool comfort even during strenuous production and maintenance procedures
  • Comfortable as under-mask protection for attachable hood veils
  • No restriction of breath or speech clarity; reduces lifting or pulling down masks
  • Will not fog safety goggles
  • More absorbent for greater particle control and longer life
Used as comfortable under-masks for hoods with built-in biological masking. They protect the wearer from contact and provide a fresh, clean barrier to the laundered mask in the hood.