Cup Style Heel Grounder, SCS, Desco Industries
Supplier: Desco
HGC1M
76238-796EA 25.59 USD
76238-796
Cup Style Heel Grounder, SCS, Desco Industries
Grounders Heel and Toe Grounders
The re-positionable configuration of the Heel Grounder extends the life of the product. An elastic loop provides a secure and comfortable fit.
  • Wide sole provides additional surface area for a better path to ground
  • Tear resistant rubber with inner scrim; reinforced material prevents tearing and increases the service life
  • Elastic hook and loop strap
  • Size adjustable material expands to fit most shoe sizes comfortably and flex during walking
  • Non-marking interior layer will not mar operator’s shoes
  • 1 megohm series resistor protects operator from accidental contact with equipment line voltage
  • Made in the United States of America
This quality Heel Grounder features a durable three-layer sole. A 24" conductive ribbon grounding tab provides additional conductive material for reliable connection with operator’s sweat-layer. Lot coded for QC traceability.
Certifications: Rg: <1.2×106 ohms per ESD SP9.2 7 ESD TR53. UL listed.