Weller® Cutting Tweezer with Narrow Oblique Heads for Soft Wires, Apex Tool Group
Supplier: Cooper Tools
15AGW
76238-602EA 167.28 USD
76238-602
Weller® Cutting Tweezer with Narrow Oblique Heads for Soft Wires, Apex Tool Group
Cutting tweezer is suitable for cutting fine, soft wires and small components.
  • Cutting tweezers with narrow oblique head
  • Soft wires up to Ø: 0.25 mm/0.010"
  • Delivers high-precision cuts
  • Hardened cutting edges for long service life
  • Suitable for cutting fine, soft wires and small components
Robust tips, non-rusting, non-reflecting surface.