Chamois® Tips Swab, ITW Chemtronics
Supplier: ITW Chemtronics
CC50
76238-532CS 158.8 USD
76238-532
Chamois® Tips Swab, ITW Chemtronics
Swabs Swabs for Clinical Use
Chamois Tips® are lint free swabs, constructed from a lint-free synthetic chamois cloth material. The chamois cloth is bonded to an ABS swab handle without potentially contaminating adhesives or glues. These swabs are ideal for cleaning and polishing magnetic and optical, audio and video heads.
  • Synthetic chamois cloth head
  • Material is free of oils and residues
  • Flexible handle bends to clean hard-to-reach areas
  • Bonded without adhesive
  • Seamless construction
Used for cleaning all optical and magnetic heads and with IPA to clean magnetic heads and drives.