Horizontal Power Beams for Arlink® 8000 Workstations, Arlink
Supplier: ARLINK DIVISION CUST SERV TE
ARL-8941
76238-456EA 407.45 USD
76238-456
Horizontal Power Beams for Arlink® 8000 Workstations, Arlink
Furniture Tables and Workbenches
Horizontal power beams for Arlink® 8000 workstations comes with eight receptacles, lighted power switch and 15 A breaker.
  • Single-sided configuration
  • Mounts in micro slots, adjusts vertically in 1" increments
Delivery information: Includes 7.5' power cord.