Pneumatic Dispenser, Ultra 850D, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
850D
76238-352EA 818.29 USD
76238-352
Pneumatic Dispenser, Ultra 850D, EFD
Fluid Adhesive Dispensers and Pistons
Fluid dispenser with analog air and vacuum control. Complete with an introductory supply of dispensing components.
Output pressure 0 to 100 psi for all fluids.