Swing Sleeves, Ansell
Supplier: Ansell Healthcare
950504
76238-262CS 257.58 USD
76238-262
Swing Sleeves, Ansell
Disposable Garments
This polyethylene sleeve is designed to protect part of the body against splashes from certain chemicals.
  • Excellent low-cost protection
  • Protects against punctures, snags and abrasions
  • Provides resistance to chemicals, fats, oils and grease
  • Elasticized sleeves opening are completely sealed with no exposed elastic
  • Heat sealed, with elastics at the extremities
Caution: The elastic band on this product may contain natural rubber latex which may cause allergic reactions in some individuals.