Syringe Barrels and Piston Sets, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7012153
76238-214BX 84.37 USD
76238-214 76241-010 76241-166 76241-722 76243-808 76245-488 76245-646 76247-468 76247-646 76252-438 76267-510
Syringe Barrels and Piston Sets, EFD
Syringe Barrels, Plungers and Pistons
Precision fit between syringe and piston ensures consistent fluid deposits.
  • Silicone-free
  • Package labels include lot numbers for process control and traceability
Wiper piston improves fluid control, keeps fluids from dripping and eliminates waste by wiping the syringe wall clean.
Sets are available in clear for most fluids, transparent amber for UV and light-sensitive materials (up to 550 nm) and opaque black for complete light blockage.
Delivery information: Each box contains one resealable bag of syringes and SmoothFlow™ pistons.
Packaging: Dust-free packaging.