Dual-Wire Thermoplastic Wrist Strap, SCS, Desco Industries
Supplier: Desco
4720
76238-086EA 29.88 USD
76238-086
Dual-Wire Thermoplastic Wrist Strap, SCS, Desco Industries
Grounders Wrist Grounders
The Dual-Wire Thermoplastic Wrist Strap consists of an insulative exterior with a conductive interior and a 3.4 mm dual conductor jack. The wrist strap is lightweight, low profile, and safely makes contact with human skin.
  • Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
  • Zipper style latching mechanism easily adjusts to most wrist sizes
  • Made in the United States of America
Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of ANSI/ESD S20.20 and Wrist Strap standard ANSI/ESD S1.1.
Ordering information: Designed for use with SCS Dual-Wire Coil Cords (sold separately).