Feather® Surgical Blade Handle, GF Health Products
Supplier: GF HEALTH PRODUCTS INC. TE
2977#3
76238-004EA 30.92 USD
76238-004
Feather® Surgical Blade Handle, GF Health Products
Scalpels and Scalpel Assemblies Scalpel Handles
The stainless steel handles are designed for smooth loading, functional handling ease and perfect balance.
  • Three handle types accommodate different size blades
  • Fits blade sizes #10, 11, 12, 15 100 mm long, flat with corrugated grip