End Cap Clamp, Holland Applied Technologies
Supplier: Holland Applied Technologies
EG16A-6L.75
76237-894EA 19.28 USD
76237-894
End Cap Clamp, Holland Applied Technologies
Fittings and Connectors
Solid end cap is made of stainless steel 304 and clamps on to the end of a process line. The end cap clamps on to the end of a ferruled tube to close the line, providing a non-permanent seal that provides take down access.
This sanitary clamp end cap meets 3-A sanitary standards and is suitable for use in food, beverage, dairy and cosmetic applications.