SnapLok™ Adapter Assemblies, Nordson EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7016129 7005011 7016127 7016059 7016122 7016135 7016113
76237-760EA 122.25 USD
76237-760 76237-762 76240-422 76241-998 76242-000 76243-746 76245-026 76250-986
SnapLok™ Adapter Assemblies, Nordson EFD
One-piece molded SnapLok™ adapter provides safe attachment of syringe barrel.
Delivery information: Includes 5/32" OØ flexible polyurethane tubing, Buna N O-ring, quick-connect and safety clip.