Human COMT ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0849
76237-646EA 1127.31 USD
76237-646
Human COMT ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human COMT AccuSignal ELISA Kit
  • Natural and recombinant human COMT.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: G52-P271
Catechol-O-methyltransferase (COMT) is one of several enzymes that degrade catecholamines such as dopamine, epinephrine, and norepinephrine. In humans, catechol-O-methyltransferase protein is encoded by the COMT gene. As the regulation of catecholamines is impaired in a number of medical conditions, several pharmaceutical drugs target COMT to alter its activity and therefore the availability of catecholamines. In the brain, COMT-dependent dopamine degradation is of particular importance in brain regions with low expression of the presynaptic dopamine transporter (DAT), such as the prefrontal cortex. This process is supposed to take place in postsynaptic neurons, as, in general, COMT is located intracellularly in the central nervous system (CNS).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human COMT standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma (heparin, EDTA) to each empty well. It is recommended that each human COMT standard solution and each sample be measured in duplicate.