Human Chemerin - RARRES2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0798
76237-624EA 760.6 USD
76237-624
Human Chemerin - RARRES2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Chemerin - RARRES2 AccuSignal ELISA Kit
  • Natural and recombinant human Chemerin.
  • There is no detectable cross-reactivity with other relevant proteins
  • Expression system for standard: E.coli; Immunogen sequence: V17-S163
Chemerin, also known as RARRES2 or TIG2, is a protein that in humans is encoded by the RARRES2 gene. It is mapped to 7q36.1. Chemerin is a potent chemoattractant specific for antigen-presenting cells that requires proteolytic activation and acts as a ligand for the G protein-coupled receptor CMKLR1(also known as ChemR23). It is a 14 kDa protein secreted in an inactive form as prochemerin and is activated through cleavage of the C-terminus by inflammatory and coagulation serine proteases. Chemerin was found to stimulate chemotaxis of dendritic cells and macrophages to the site of inflammation. What’s more, the active protein has several roles, including that as an adipokine, and is truncated on both termini from the proprotein.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human Chemerin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human Chemerin standard solution and each sample is measured in duplicate.