Human IFN gamma ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0190
76237-584EA 679.1 USD
76237-584
Human IFN gamma ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human IFN gamma AccuSignal ELISA Kit
  • Natural and recombinant human IFN gamma.
  • Expression system for standard: E.coli; Immunogen sequence: Q24-Q166
Interferon-gamma(IFN-gamma) is an inflammatory cytokine that has been implicated in the development of fibrosis in inflamed tissues. The production of IFN-gamma, which is under genetic control, can influence the development of fibrosis in lung allografts. IFN-gamma is also produced by natural killer(NK) cells and most prominently by CD8 cytotoxic T cells, and is vital for the control of microbial pathogens. Interferon gamma is believed to be crucial for host defence against many infections. Genetically determined variability in IFN-gamma and expression might be important for the development of tuberculosis. IFN-gamma activates human macrophage oxidative metabolism and antimicrobial activity. In addition to having antiviral activity, IFN-gamma has important immunoregulatory functions. IFN-gamma plays an important role in the control of neointima proliferation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human IFN gamma standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human IFN gamma standard solution and each sample be measured in duplicate.