Rat GDNF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0181
76237-566EA 760.6 USD
76237-566
Rat GDNF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat GDNF AccuSignal ELISA Kit
  • Natural and recombinant rat GDNF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: S78-I211
Glial cell line-derived neurotrophic factor(GDNF) is a glycosylated, disulfide-bonded homodimer that is a distantly related member of the transforming growth factor-beta superfamily. GDNF, is a potent neurotrophic factor that promotes the survival of dopaminergic neurones in cultures including embryonic neuronal cultures. GDNF, in addition to its potential role in the differentiation and survival of central nervous system neurons, has profound effects on kidney organogenesis and the development of the peripheral nervous system.3 GDNF may have utility in the treatment of Parkinson's disease, which is marked by progressive degeneration of midbrain dopaminergic neurons. GDNF lies on the short arm of human chromosome 5, at 5p13.1-p13.3 ability to promote dopamine uptake in midbrain cultures. The standard product used in this kit is recombinant rat GDNF, which is a dimmer composed of two chains with 134 amino acids.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat GDNF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each rat GDNF standard solution and each sample is measured in duplicate.