Human G-CSF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0178
76237-560EA 760.6 USD
76237-560
Human G-CSF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human G-CSF AccuSignal ELISA Kit
  • Natural and recombinant human G-CSF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: T31-P204
Granulocyte colony-stimulating factor(G-CSF) is a member of the CSF family of hormone-like glycoprotein that regulates hematopoietic cell proliferation and differentiation, and it almost exclusively stimulates the colony formation of granulocytes from committed precursor cells in semi-solid agar culture. G-CSF is also termed colony stimulating factor-3, and a single gene of which codes for a 177 or 180 amino acid mature protein of molecular weight 19,600. Functionally, it specifically stimulates the proliferation and differentiation of the progenitor cells for granulocytes. The effect of G-CSF on myeloid leukemias is unique among colony stimulating factors in driving the leukemic cells from a self-renewing malignant state to a mature differentiated phenotype with the concomitant loss of tumorigenicity. Besides, it also prevents cardiac remodeling after myocardial infarction by activating the Jak-Stat pathway in cardiomyocytes. The recombinant form of hG-CSF is capable of supporting neutrophil proliferation in a CFU-GM assay as well as early erythroid colonies and mixed colony formation. Human gene coding for G-CSF is assigned to the q21-q22 region of chromosome 17. The standard product used in this kit is recombinant human G-CSF, consisting of 175 amino acids with the molecular mass of 18.8KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human G-CSF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human G-CSF standard solution and each sample be measured in duplicate.