Borate Saline Buffered (BSB), Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-051-1000
76237-552EA 92.99 USD
76237-552
Borate Saline Buffered (BSB), Rockland Immunochemicals
Buffers Electrophoresis Buffers Blocking Buffers
Borate Saline Buffered (BSB) is commonly used in coating procedures for ELISA.
  • Borate Saline Buffered - BSB
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
Borate Saline Buffered (BSB) is commonly used in coating procedures for ELISA. BSB can also be used as a component in blocking solutions for Western blotting. The buffer can be used as a diluent in biological samples and for various applications within biology and biochemistry to maintain the pH.
Application Note: Borate Saline Buffered (BSB) is commonly used in coating procedures for ELISA. BBS can also be used as a component in blocking solutions for Western blotting. The buffer can be used as a diluent in biological samples and for various applications within biology and biochemistry to maintain the pH.