Borate Buffered Saline (BBS), Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-050-1000
76237-550EA 92.99 USD
76237-550
Borate Buffered Saline (BBS), Rockland Immunochemicals
Buffers Electrophoresis Buffers Blocking Buffers
Borate buffered saline (BBS) is commonly used in coating procedures for ELISA.
  • Borate buffered saline - BBS
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
BBS can also be used as a component in blocking solutions for Western blotting. The buffer can be used as a diluent in biological samples and for various applications within biology and biochemistry to maintain the pH.
Application Note: BBS is isotonic and has a strong bactericidal effect. It can be used to dilute solutions for coating procedures. Additives such as Tween 20 and Blotto can be used to add functionality as a washing buffer or blocking buffer.