Human Syndecan-4 - SDC4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Human Syndecan-4 - SDC4 AccuSignal ELISA Kit
- Natural and recombinant human Syndecan-4.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E19-E145
Syndecan-4 (SDC4) is a protein that in humans is encoded by the SDC4 gene. The protein is found as a homodimer and is a member of the syndecan proteoglycan family. It is mapped to 20q13.12. Syndecan-4 is one of the four vertebrate syndecans and has a molecular weight of ~20 kDa. It is a transmembrane (type I) heparan sulfate proteoglycan that functions as a receptor in intracellular signaling. Syndecan-4 also regulates the actin cytoskeleton, cell adhesion, and cell migration. In addition, Syndecan-4 can activate protein kinase C (PKC). The variable domain of syndecan-4 could be a site of self-association. What’s more, Syndecan-4 also binds to phosphatidylinositol (4,5)-bisphosphate (PIP2) through the variable domain and increases PKC activity ten-fold.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Syndecan-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human Syndecan-4 standard solution and each sample be measured in duplicate.