Human CCL16 - HCC-4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0645
76237-456EA 760.6 USD
76237-456
Human CCL16 - HCC-4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL16 - HCC-4 AccuSignal ELISA Kit
  • Natural and recombinant human CCL16.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q24-Q120
Chemokine(C-C motif) ligand 16(CCL16) is a small cytokine belonging to the CC chemokine family that is known under several pseudonyms, including Liver-expressed chemokine(LEC) and Monotactin-1(MTN-1). This chemokine is expressed by the liver, thymus, and spleen and is chemoattractive for monocytes and lymphocytes. Cellular expression of CCL16 can be strongly induced in monocytes by IL-10, IFN-gamma and bacterial lipopolysaccharide. Its gene is located on chromosome 17, in humans, among a cluster of other CC chemokines. CCL16 elicits its effects on cells by interacting with cell surface chemokine receptors such as CCR1, CCR2, CCR5 and CCR8.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human CCL16 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human CCL16 standard solution and each sample is measured in duplicate.