Human CD244 - 2B4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0562
76237-412EA 760.6 USD
76237-412
Human CD244 - 2B4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD244 - 2B4 AccuSignal ELISA Kit
  • Natural and recombinant human CD244.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: C22-R221
CD244(Cluster of Differentiation 244) is a human protein encoded by the CD244 gene. It is also known as Natural Killer Cell Receptor 2B4. Members of the CD2 family adhere to each other instead,the cell surface glycoprotein 2B4 is related to CD2 and is implicated in the regulation of NK- and T-cell function. The 2B4 gene was mapped to 1q22 on gene map, where SLAM, CD48, CD84 and LY9 are also located. Using recombinant human NK cell-activating ligand 2B4 fused to domains 3 and 4 of rodent Cd4 and flow cytometric analysis, It was found that CD48 binds to 2B4. BIAcore surface plasmon resonance analysis showed that the affinity of CD48 for 2B4 is approximately 10-fold higher than that shown for CD48 and CD2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human CD244 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD244 standard solution and each sample be measured in duplicate.