Human IL-17RA ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0463
76237-312EA 760.6 USD
76237-312
Human IL-17RA ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-17RA AccuSignal ELISA Kit
  • Natural and recombinant human IL-17RA.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L33-W320
Interleukin 17 receptor A, also known as IL17RA and CDw217 (cluster of differentiation w217), is a human gene. The IL17R gene is mapped to 22q11.22-q11.23 by radiation hybrid analysis. Interleukin 17A (IL17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. The protein encoded by this gene is a ubiquitous type I membrane glycoprotein that binds with low affinity to interleukin 17A. Interleukin 17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Like other cytokine receptors, this receptor likely has a multimeric structure. Also, it is showed that mouse Il17r bound HSV-13 and mouse Il17. Recombinant soluble Il17r inhibited mitogen-induced T-cell proliferation and Il2 production.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human IL-17RA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma (heparin, EDTA) to each empty well. It is recommended that each human IL-17RA standard solution and each sample be measured in duplicate.