Human Adiponectin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0365
76237-196EA 777.31 USD
76237-196
Human Adiponectin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Adiponectin AccuSignal ELISA Kit
  • Natural and recombinant human total Adiponectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E19-N244
Adiponectin(ADPN) is a hormone secreted by adipocytes that regulates energy homeostasis and glucose and lipid metabolism. Adiponectin is a new member of the family of soluble defense collagens, in hematopoiesis and immune responses. It is an important negative regulator in hematopoiesis and immune systems and raise the possibility that it may be involved in ending inflammatory responses through its inhibitory functions. Adiponectin is mapped to 3q27 and can protect the organism from systemic inflammation by promoting the clearance of early apoptotic cells by macrophages through a receptor-dependent pathway involving calreticulin. The standard product used in this kit is the product of gene recombination, consisting of 226(19-244) amino acids with the molecular mass of 36KDa after glycosylation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 100ng/ml, 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml human Adiponectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), tissue homogenates or urine to each empty well. It is recommended that each human Adiponectin standard solution and each sample be measured in duplicate.