Human SCF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0303
76237-184EA 760.6 USD
76237-184
Human SCF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human SCF AccuSignal ELISA Kit
  • Natural and recombinant human SCF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: E26-A190
Stem Cell Factor (also known as SCF, kit-ligand, KL, or steel factor) is a cytokine that binds to the c-Kit receptor (CD117). It is mapped to 12q21.32. SCF was primarily expressed by perivascular cells throughout the bone marrow, and it plays a role in the regulation of HSCs in the stem cell niche in the bone marrow. SCF can exist both as a transmembrane protein and a soluble protein. This gene plays an important role in the hematopoiesis during embryonic development. During development, the presence of the SCF also plays an important role in the localization of melanocytes, cells that produce melanin and control pigmentation. In addition to it, SCF can promote mast cell adhesion, migration, proliferation, and survival. It also promotes the release of histamine and tryptase, which are involved in the allergic response.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human SCF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA, citrate) to each empty well. It is recommended that each human SCF standard solution and each sample be measured in duplicate.