Human Eotaxin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0159
76237-130EA 760.6 USD
76237-130
Human Eotaxin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Eotaxin AccuSignal ELISA Kit
  • Natural and recombinant human Eotaxin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: G24-P97
Eotaxin, also known as CCL11, is a potent inducer of eosinophil chemotaxis and is considered as a selective ligand of the CC chemokine receptor 3(CCR3), which is expressed on eosinophils, basophils, and Th2 lymphocytes. Eotaxin is assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections. The gene maps to chromosome 17 and is expressed constitutively at high levels in small intestine and colon, and at lower levels in various other tissues. The deduced mature protein sequence is 66% identical to human monocyte chemoattractant protein-1, and 60% identical to guinea pig eotaxin. Recombinant human eotaxin produced in insect cells induces a calcium flux response in normal human eosinophils, but not in neutrophils or monocytes. The human eotaxin gene is cloned and found to be 61.8 and 63.2% identical at the amino acid level to guinea pig and mouse eotaxin.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human Eotaxin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(citrate,EDTA) to each empty well. It is recommended that each human Eotaxin standard solution and each sample be measured in duplicate.