QuantiChrom™ Peroxidase Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
D2PD-100
76237-096EA 495.47 USD
76237-096
QuantiChrom™ Peroxidase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of peroxidase activity.
  • Fast and sensitive, convenient and high-throughput
  • Use as little as 10 μl samples
  • Linear detection range: colorimetric assays 2 to 50 U/l, fluorimetric assays 0.1 to 5 U/l peroxidase
  • Homogeneous "mix-incubate-measure" type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: OD570 nm, or FL530/585 nm
  • Samples: Biological
Peroxidase (EC number 1.11.1.x) catalyze the following oxidation-reduction reactions:
ROOR + electron donor (2 e-) + 2H+ → ROH + R’OH. For many peroxidases the optimal substrate is hydrogen peroxide (H₂O₂), but others are more active with organic hydroperoxides such as lipid peroxides. In the cell, peroxidases destroy toxic hydroxide radicals that are formed as byproducts during aerobic respiration. The peroxidases represent a large family of enzymes that are found in animals (e.g. myeloperoxidase-like enzymes), plant, fungi and bacteria (cytochrome-c peroxidase like enzymes such a horseradish peroxidase). Simple, direct and automation-ready procedures for determining peroxidase activity find wide applications. Peroxidase assay uses H₂O₂ and an electron donor dye that forms a pink color during the peroxidase reaction. The optical density (570nm) or fluorescence intensity (λex/em = 530/590nm) is a direct measure of the enzyme activity.