Human TNFSF4 - OX40L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0518
76236-924EA 760.6 USD
76236-924
Human TNFSF4 - OX40L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TNFSF4 - OX40L AccuSignal ELISA Kit
  • Natural and recombinant human TNFSF4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q51-L183
OX40L is the ligand for CD134 and is expressed on such cells as DC2s enabling amplification of Th2 cell differentiation. OX40L has also been designated CD252. By analysis of an interspecific backcross, the mouse Tnfsf4 gene is mapped to chromosome 1. Using fluorescence in situ hybridization, they localized the human TNFSF4 gene to 1q25, a region sharing homology of synteny with the portion of mouse chromosome 1 containing the Tnfsf4 gene. OX40L strongly inhibited the generation of IL-10-producing Tr1 cells induced by two physiologic stimuli, the inducible costimulatory ligand and immature dendritic cells. In addition, OX40L strongly inhibited IL-10 production and suppressive function of differentiated IL-10-producing Tr1 cells. Tnfsf4 underlies Ath1 in mice and that polymorphisms in its human homolog TNFSF4 increase the risk of myocardial infarction in humans.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml human TNFSF4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each human TNFSF4 standard solution and each sample be measured in duplicate.